Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243132.1 | NP_001230061.1 | 8 | 1078 | ||
NM_027533.3 | NP_081809.2 | 8 | 1273 | |||
GenBank mRNA | AK018178.1 | - | 8 | 1076 | ||
AK031683.1 | - | 7 | 983 | |||
AK032520.1 | - | 8 | 1099 | |||
AK049888.1 | - | 8 | 1075 | |||
AK079560.1 | - | 8 | 1077 | |||
AK160479.1 | - | 1 | 357 | |||
BC007185.1 | - | 8 | 1513 | 3591472 |