Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027733.5 | NP_082009.3 | 3 | 294 | ||
NM_029485.2 | NP_083761.1 | 3 | 294 | |||
GenBank mRNA | AK016341.1 | - | 3 | 300 | ||
AK017218.1 | - | 3 | 270 | |||
AY730532.1 | - | 3 | 294 | |||
BC060950.1 | - | 3 | 247 | 6773733 | ||
BU961109.1 | - | 3 | 297 |