Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027817.3 | NP_082093.1 | 2 | 169 | ||
GenBank mRNA | AK018457.1 | - | 2 | 170 | ||
AK153888.1 | - | 2 | 226 | |||
BC120674.1 | - | 2 | 164 | 40129597 | ||
BC120676.1 | - | 2 | 164 | 40129599 |