Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000169.2 | NP_000160.1 | 6 | 1060 | ||
GenBank mRNA | AK222627.1 | - | 6 | 972 | ||
AK291095.1 | - | 6 | 979 | |||
BC002689.2 | - | 6 | 956 | 3609235 | ||
BT006864.1 | - | 6 | 950 | |||
D00039.1 | - | 6 | 874 | |||
X05790.1 | - | 6 | 973 | |||
X16889.1 | - | 6 | 952 |