Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027629.3 | NP_081905.1 | 2 | 560 | ||
GenBank mRNA | AK039189.1 | - | 2 | 544 | ||
AK040513.1 | - | 2 | 480 | |||
BB633030.1 | - | 2 | 541 | |||
BC020351.1 | - | 2 | 773 | |||
BC076571.1 | - | 2 | 457 | 30356163 | ||
CF531609.1 | - | 2 | 457 |