Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027650.3 | NP_081926.2 | 5 | 989 | ||
GenBank mRNA | AK016676.1 | - | 5 | 976 |
Gene Symbol: Speer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173069.3 | NP_775092.3 | 6 | 1091 | ||
GenBank mRNA | AV044901.3 | - | 3 | 556 |