Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027865.2 | NP_082141.1 | 5 | 853 | ||
GenBank mRNA | AK002841.1 | - | 5 | 854 | ||
AK154206.1 | - | 4 | 1339 | |||
AK165199.1 | - | 5 | 885 | |||
AK165827.1 | - | 5 | 838 | |||
BC060057.1 | - | 5 | 873 | 5696102 | ||
BC060243.1 | - | 5 | 873 | 5696102 |