Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163645.1 | NP_001157117.1 | 22 | 3064 | ||
NM_027881.3 | NP_082157.2 | 22 | 2782 | |||
GenBank mRNA | AK004768.1 | - | 22 | 2784 | ||
AK042104.1 | - | 2 | 1161 | |||
BC044866.1 | - | 21 | 2709 | 5003631 | ||
BC086485.1 | - | 22 | 2976 | 5344992 | ||
CD538704.1 | - | 1 | 115 | |||
CF751761.1 | - | 3 | 331 |