Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027997.2 | NP_082273.1 | 2 | 263 | ||
GenBank mRNA | AK009343.1 | - | 2 | 263 | ||
BC132287.1 | - | 2 | 250 | 40130564 | ||
BC144769.1 | - | 2 | 251 | 9053317 |