Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000210.3 | NP_000201.2 | 11 | 1736 | ||
NM_001079818.2 | NP_001073286.1 | 11 | 1736 | |||
NM_001316306.1 | NP_001303235.1 | 11 | 1422 | |||
XM_006712510.1 | XP_006712573.1 | 11 | 1705 | |||
XM_006712511.1 | XP_006712574.1 | 11 | 1705 | |||
XM_017004005.1 | XP_016859494.1 | 11 | 1432 | |||
XM_017004006.1 | XP_016859495.1 | 11 | 1432 | |||
XM_017004007.1 | XP_016859496.1 | 11 | 1432 | |||
XM_017004008.1 | XP_016859497.1 | 11 | 1432 | |||
GenBank mRNA | AB208842.1 | - | 11 | 1404 | ||
AK294436.1 | - | 7 | 824 | |||
AK296496.1 | - | 11 | 1358 | |||
BC007002.1 | - | 11 | 1702 | |||
BC050585.1 | - | 5 | 387 | |||
BC136455.1 | - | 11 | 1610 | 9020444 | ||
BC136456.1 | - | 11 | 1610 | 9020445 | ||
X53586.1 | - | 11 | 1648 | |||
X59512.1 | - | 11 | 1484 |