Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039088.1 | NP_001034177.1 | 1, 2 | 253 | ||
NM_028112.2 | NP_082388.1 | 1, 2 | 253 | |||
GenBank mRNA | AK011330.1 | - | 1, 2 | 155 | ||
AK088835.1 | - | 1, 2 | 127 | |||
AK143727.1 | - | 1, 2 | 253 | |||
AK169231.1 | - | 1, 2 | 134 | |||
AK169388.1 | - | 1, 2 | 133 | |||
AK170591.1 | - | 1, 2 | 206 | |||
BC027244.1 | - | 1, 2 | 205 | 3584697 | ||
BY732540.1 | - | 1, 2 | 253 |