Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027926.2 | NP_082202.1 | 6, 7 | 610 | ||
GenBank mRNA | AK030705.1 | - | 6, 7 | 610 | ||
BC061206.1 | - | 6, 7 | 617 | 30249166 |