Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028015.2 | NP_082291.1 | 10 | 1748 | ||
GenBank mRNA | AK010241.1 | - | 10 | 1676 | ||
AK145589.1 | - | 10 | 1656 | |||
AK145762.1 | - | 10 | 1670 | |||
AK146220.1 | - | 10 | 1670 | |||
AK146417.1 | - | 10 | 1640 | |||
AK208035.1 | - | 1 | 127 | |||
AY029533.1 | - | 10 | 1668 | |||
BC010670.1 | - | 4 | 969 | |||
BC043059.1 | - | 1802 | 6419704 | |||
BC046797.1 | - | 10 | 1748 | 6403129 |