Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028087.2 | NP_082363.2 | 3 | 1762 | ||
GenBank mRNA | AK008234.1 | - | 2 | 1372 | ||
AY039027.1 | - | 2 | 1436 | |||
BC018297.1 | - | 2 | 1324 | 4208500 | ||
EF202835.1 | - | 3 | 1762 |