Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285839.1 | NP_001272768.1 | 4 | 1067 | ||
GenBank mRNA | AK011265.1 | - | 4 | 1015 | ||
AK045669.1 | - | 4 | 1029 | |||
AK137332.1 | - | 4 | 1018 | |||
BC030671.1 | - | 4 | 1032 | |||
BC038910.1 | - | 2 | 393 | |||
BC051211.1 | - | 1 | 443 | |||
BC058172.1 | - | 4 | 1030 | 5012054 |