Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028115.3 | NP_082391.1 | 3 | 416 | ||
NM_029839.3 | NP_084115.2 | 3 | 416 | |||
GenBank mRNA | AK011362.1 | - | 3 | 411 | ||
AK018525.1 | - | 3 | 389 | |||
AK031674.1 | - | 3 | 394 | |||
AK164095.1 | - | 3 | 387 | |||
AK169221.1 | - | 3 | 382 | |||
BC039287.1 | - | 3 | 361 | |||
BC054088.1 | - | 3 | 230 | |||
BC089592.1 | - | 3 | 383 | 6757213 |