Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011832.1 | - | 3 | 191 | ||
AK030398.1 | - | 3 | 390 | |||
AK047429.1 | - | 2 | 173 | |||
AK134257.1 | - | 3 | 190 | |||
AK135723.1 | - | 2 | 174 | |||
AK162903.1 | - | 2 | 613 | |||
AY092766.1 | - | 2 | 282 | |||
BC023762.1 | - | 3 | 460 | |||
BC057991.1 | - | 3 | 231 | 5323559 | ||
BE994245.1 | - | 2 | 332 | |||
EF410128.1 | - | 415 |