Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 2, 3 | 413 | ||
GenBank mRNA | AF512670.1 | - | 2, 3 | 119 | ||
AF536771.1 | - | 2, 3 | 182 | |||
AK013600.1 | - | 2, 3 | 415 | |||
AK018119.1 | - | 2, 3 | 370 | |||
AK039592.1 | - | 2, 3 | 256 | |||
AK079059.1 | - | 863 | ||||
AK148956.1 | - | 2, 3 | 384 | |||
BC026686.1 | - | 2, 3 | 351 | 5065516 | ||
BC066007.1 | - | 2, 3 | 653 | |||
BC085147.1 | - | 2, 3 | 565 | |||
BC088979.1 | - | 2, 3 | 348 | 5690600 |