Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028390.3 | NP_082666.2 | 4 | 986 | ||
GenBank mRNA | AK013624.1 | - | 1 | 30 | ||
AK038403.1 | - | 1 | 61 | |||
BC027141.1 | - | 4 | 950 | |||
BC032164.1 | - | 4 | 1004 | 5346703 |