Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 4 | 633 | ||
GenBank mRNA | AF512670.1 | - | 4 | 339 | ||
AF536771.1 | - | 4 | 402 | |||
AK013600.1 | - | 4 | 635 | |||
AK018119.1 | - | 4 | 590 | |||
AK039592.1 | - | 4 | 476 | |||
AK079059.1 | - | 1083 | ||||
AK148956.1 | - | 4 | 604 | |||
BC026686.1 | - | 4 | 571 | 5065516 | ||
BC066007.1 | - | 4 | 873 | |||
BC085147.1 | - | 4 | 786 | |||
BC088979.1 | - | 4 | 568 | 5690600 |