Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000282.3 | NP_000273.2 | 23 | 2164 | ||
NM_001127692.2 | NP_001121164.1 | 22 | 2086 | |||
NM_001178004.1 | NP_001171475.1 | 22 | 2023 | |||
XM_017020608.1 | XP_016876097.1 | 22 | 2105 | |||
XM_017020609.1 | XP_016876098.1 | 23 | 2072 | |||
XM_017020613.1 | XP_016876102.1 | 23 | 2097 | |||
XM_017020614.1 | XP_016876103.1 | 21 | 1869 | |||
XM_017020617.1 | XP_016876106.1 | 19 | 1743 | |||
XM_017020618.1 | XP_016876107.1 | 18 | 1689 | |||
GenBank mRNA | AF385926.1 | - | 23 | 2096 | ||
AK225288.1 | - | 23 | 2064 | |||
AK225299.1 | - | 23 | 2062 | |||
AK296771.1 | - | 22 | 1984 | |||
AK298318.1 | - | 22 | 1945 | |||
AK315503.1 | - | 23 | 2062 | |||
BC000140.2 | - | 23 | 2036 | 3048998 | ||
M26121.1 | - | 2 | 162 | |||
S79219.1 | - | 7 | 556 | |||
X14608.1 | - | 23 | 2031 |