Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172152.1 | NP_001165623.1 | 8, 9 | 1362 | ||
NM_028220.3 | NP_082496.3 | 9, 10 | 1385 | |||
GenBank mRNA | AK044241.1 | - | 8, 9 | 1363 | ||
BC028268.1 | - | 9, 10 | 1396 | 5366098 |