Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028279.3 | NP_082555.2 | 17 | 1955 | ||
GenBank mRNA | AK012270.1 | - | 4 | 530 | ||
AK133611.1 | - | 17 | 2065 | |||
AK135653.1 | - | 19 | 2098 | |||
AK161457.1 | - | 20 | 2261 | |||
AY243507.1 | - | 17 | 1955 | |||
BC108995.1 | - | 17 | 1939 | 40047859 | ||
BC108996.1 | - | 17 | 1939 | 40047860 |