Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130879.2 | NP_570949.2 | 28 | 5372 | ||
GenBank mRNA | AK013309.1 | - | 1 | 1128 | ||
BC019125.1 | - | 1 | 589 | |||
BC058410.1 | - | 11 | 3018 | |||
BC085481.1 | - | 1 | 1619 | |||
BC108359.1 | - | 1 | 1651 |