Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 9 | 1943 | ||
GenBank mRNA | AK013600.1 | - | 9 | 1945 | ||
AK018119.1 | - | 9 | 1906 | |||
AK039592.1 | - | 9 | 1787 | |||
AK148956.1 | - | 9 | 1914 | |||
BB694259.1 | - | 1 | 306 | |||
BC026686.1 | - | 9 | 1881 | 5065516 | ||
BC066007.1 | - | 9 | 2184 | |||
BC085147.1 | - | 9 | 2096 | |||
BC088979.1 | - | 9 | 1878 | 5690600 |