Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028416.2 | NP_082692.1 | 8 | 1185 | ||
GenBank mRNA | AJ457192.1 | - | 8 | 1197 | ||
AK013685.1 | - | 2 | 96 | |||
BC120532.1 | - | 8 | 1128 | 40129455 |