Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700029M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006510.2 | - | 3 | 530 | ||
AK015107.1 | - | 2 | 353 | |||
AK016438.1 | - | 3 | 647 | |||
BC059720.1 | - | 2 | 375 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_028688.1 | - | 353 | Chr. - - [-] on Build |