Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000326.4 | NP_000317.1 | 4 | 472 | ||
XM_011521870.2 | XP_011520172.1 | 4 | 1901 | |||
XM_017022460.1 | XP_016877949.1 | 1 | 311 | |||
GenBank mRNA | AK312457.1 | - | 4 | 360 | ||
BC004199.2 | - | 4 | 374 | 3604048 | ||
BM921294.1 | - | 4 | 541 | |||
BX490622.1 | - | 4 | 473 | |||
J04213.1 | - | 3 | 275 |