Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028724.4 | NP_083000.5 | 9 | 1735 | ||
GenBank mRNA | AK014548.1 | - | 1 | 207 | ||
AK040763.1 | - | 1 | 207 | |||
AK150680.1 | - | 7 | 1543 | |||
AK151332.1 | - | 6 | 1499 | |||
AK152025.1 | - | 1 | 180 | |||
AK152893.1 | - | 7 | 1543 | |||
AK154421.1 | - | 1 | 151 | |||
AK163047.1 | - | 1 | 722 | |||
AK171064.1 | - | 1 | 122 | |||
BC005529.1 | - | 1 | 168 | 3495681 | ||
BC040390.1 | - | 9 | 1729 | 3156682 |