Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028524.3 | NP_082800.1 | 4 | 612 | ||
GenBank mRNA | AK005970.1 | - | 4 | 606 | ||
AY162282.1 | - | 4 | 492 | |||
AY241872.1 | - | 4 | 606 | |||
BC050983.1 | - | 4 | 609 | |||
BC060997.1 | - | 4 | 505 | 6742828 |