Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110824.1 | NP_001104294.1 | 5 | 899 | ||
NM_001110825.1 | NP_001104295.1 | 5 | 899 | |||
NM_028767.2 | NP_083043.2 | 5 | 899 | |||
GenBank mRNA | AB052766.1 | - | 5 | 733 | ||
AK004693.1 | - | 5 | 710 | |||
AK143194.1 | - | 5 | 744 | |||
AY135029.1 | - | 5 | 687 | |||
BC023368.1 | - | 4 | 1156 | |||
BC043702.1 | - | 5 | 673 | 5351757 | ||
BC052407.1 | - | 5 | 702 | 5716170 | ||
BC057110.1 | - | 5 | 899 | 6417088 |