Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ampd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289719.1 | NP_001276648.1 | 3, 4 | 490 | ||
NM_001289720.1 | NP_001276649.1 | 2, 3 | 307 | |||
NM_028779.5 | NP_083055.1 | 2, 3 | 576 | |||
GenBank mRNA | AK004759.2 | - | 3, 4 | 475 | ||
AK034844.1 | - | 2, 3 | 280 | |||
AK169980.1 | - | 3, 4 | 476 | |||
BC049119.1 | - | 2, 3 | 537 | 6812571 | ||
BY209955.1 | - | 307 |