Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028772.3 | NP_083048.1 | 2 | 208 | ||
GenBank mRNA | AK004755.1 | - | 2 | 209 | ||
BC089599.1 | - | 2 | 211 | 30118948 |