Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acoxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028765.3 | NP_083041.1 | 10 | 1108 | ||
GenBank mRNA | AK004775.1 | - | 10 | 1110 | ||
BC048717.1 | - | 3 | 204 | 6334753 |