Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110824.1 | NP_001104294.1 | 12, 13 | 1901 | ||
NM_001110825.1 | NP_001104295.1 | 12, 13 | 1865 | |||
NM_028767.2 | NP_083043.2 | 12, 13 | 1862 | |||
GenBank mRNA | AB052766.1 | - | 12, 13 | 1696 | ||
AK004693.1 | - | 12, 13 | 1673 | |||
AK143194.1 | - | 12, 13 | 1712 | |||
AY135029.1 | - | 12, 13 | 1689 | |||
BC043702.1 | - | 12, 13 | 1639 | 5351757 | ||
BC052407.1 | - | 12, 13 | 1704 | 5716170 | ||
BC057110.1 | - | 12, 13 | 1862 | 6417088 |