Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005891.1 | - | 5 | 2473 | ||
BC060685.1 | - | 4 | 4931 | |||
BC062897.1 | - | 5 | 3397 |