Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028868.3 | NP_083144.1 | 15 | 2142 | ||
GenBank mRNA | AK010337.1 | - | 15 | 2143 | ||
AK083655.1 | - | 15 | 2147 | |||
AK146688.1 | - | 15 | 2226 | |||
AY099096.1 | - | 15 | 2114 | |||
BC012261.1 | - | 11 | 1685 | |||
BC030938.1 | - | 15 | 1974 | 3980339 |