Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029203.2 | NP_083479.1 | 2 | 199 | ||
GenBank mRNA | AK015997.1 | - | 2 | 200 | ||
BC132216.1 | - | 2 | 193 | 40130493 | ||
BC132220.1 | - | 2 | 193 | 40130497 | ||
BC145534.1 | - | 2 | 193 | |||
DQ058639.1 | - | 2 | 200 |
Gene Symbol: Rhox2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099316.1 | NP_001092786.1 | 2 | 177 | ||
GenBank mRNA | BC049546.1 | - | 177 |
Gene Symbol: Rhox2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085348.1 | NP_001078817.1 | 199 |
Gene Symbol: Rhox2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085356.1 | NP_001078825.1 | 2 | 199 |