Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029115.3 | NP_083391.2 | 3 | 511 | ||
GenBank mRNA | AK015464.1 | - | 3 | 498 | ||
AK076640.1 | - | 3 | 506 | |||
AK149168.1 | - | 3 | 506 | |||
BC048086.1 | - | 3 | 403 | 6390120 | ||
BC060044.1 | - | 3 | 510 |