Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028990.4 | NP_083266.1 | 5 | 2475 | ||
GenBank mRNA | AK018471.1 | - | 4 | 1205 | ||
AK034589.1 | - | 5 | 2444 | |||
AK051532.1 | - | 5 | 2476 | |||
AK054353.1 | - | 5 | 2432 | |||
AK087647.1 | - | 1 | 86 | |||
AK152957.1 | - | 5 | 2438 | |||
AK170302.1 | - | 5 | 2452 | |||
BC007160.1 | - | 5 | 2419 | 3488466 | ||
BC024911.1 | - | 5 | 2394 | 3594306 |