Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029037.4 | NP_083313.1 | 5 | 3561 | ||
GenBank mRNA | AK043620.1 | - | 5 | 3562 |