Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029101.4 | NP_083377.3 | 7 | 1935 | ||
GenBank mRNA | AK003698.1 | - | 7 | 1886 | ||
AK032596.1 | - | 1898 | ||||
AK036247.1 | - | 1880 | ||||
AK088456.1 | - | 7 | 1914 | |||
AK146821.1 | - | 7 | 1898 | |||
AK153565.1 | - | 7 | 1894 | |||
AK159553.1 | - | 7 | 1880 | |||
AK168850.1 | - | 7 | 1888 | |||
AK189914.1 | - | 1 | 106 | |||
AK205996.1 | - | 1 | 106 | |||
AK212057.1 | - | 1 | 106 | |||
BC012523.1 | - | 7 | 1872 | 3602183 |