Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rhox2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099318.1 | NP_001092788.1 | 561 |
Gene Symbol: Rhox2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029203.2 | NP_083479.1 | 3, 4 | 561 | ||
GenBank mRNA | AK015997.1 | - | 3, 4 | 562 | ||
BC132216.1 | - | 3, 4 | 555 | 40130493 | ||
BC132220.1 | - | 3, 4 | 555 | 40130497 | ||
DQ058639.1 | - | 3, 4 | 562 |
Gene Symbol: Rhox2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099316.1 | NP_001092786.1 | 3, 4 | 539 | ||
GenBank mRNA | BC049546.1 | - | 539 |
Gene Symbol: Rhox2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081669.2 | NP_001075138.2 | 3, 4 | 572 |
Gene Symbol: Rhox2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085348.1 | NP_001078817.1 | 561 |
Gene Symbol: Rhox2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085356.1 | NP_001078825.1 | 3, 4 | 561 |
Gene Symbol: Rhox2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114153.1 | NP_001107625.1 | 3, 4 | 577 |
Gene Symbol: Rhox2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100465.1 | NP_001093935.1 | 3, 4 | 561 |