Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000423.2 | NP_000414.2 | 2 | 653 | ||
GenBank mRNA | BC096294.4 | - | 2 | 642 | 40007308 | |
BC099643.1 | - | 2 | 642 | 40007309 | ||
BC099644.3 | - | 2 | 642 | 40007310 | ||
M99061.1 | - | 2 | 671 |