Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029457.3 | NP_083733.1 | 2, 3 | 315 | ||
GenBank mRNA | AK015987.1 | - | 2, 3 | 294 | ||
AK031030.1 | - | 2, 3 | 300 | |||
BC031652.1 | - | 2, 3 | 290 | 3483046 | ||
CJ169147.1 | - | 2, 3 | 315 |