Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029576.3 | NP_083852.1 | 4 | 278 | ||
GenBank mRNA | AB232584.1 | - | 3 | 220 | ||
AK003609.1 | - | 4 | 279 | |||
AK042020.1 | - | 4 | 280 | |||
AK156726.1 | - | 4 | 301 | |||
BC016408.1 | - | 4 | 273 | 4242206 |