Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029576.3 | NP_083852.1 | 6 | 546 | ||
GenBank mRNA | AB232584.1 | - | 5 | 488 | ||
AK003609.1 | - | 6 | 547 | |||
AK042020.1 | - | 6 | 548 | |||
AK156726.1 | - | 6 | 569 | |||
AK200798.1 | - | 2 | 195 | |||
BC016408.1 | - | 6 | 541 | 4242206 |