Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
PLEC
Gene Name:
plectin
Gene Aliases:
EBS1, EBSMD, EBSND, EBSO, EBSOG, EBSPA, HD1, LGMD2Q, PCN, PLEC1, PLEC1b, PLTN
Chromosome Location:
Chr.8: 143915147 - 143976800 on Build GRCh38
UniGene ID:
Hs.434248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
8395
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000445.4
NP_000436.2
4
320
NM_201378.3
NP_958780.1
3
205
NM_201379.2
NP_958781.1
3
273
NM_201380.3
NP_958782.1
3
771
NM_201381.2
NP_958783.1
3
168
NM_201382.3
NP_958784.1
3
258
NM_201383.2
NP_958785.1
3
211
NM_201384.2
NP_958786.1
3
319
XM_005250976.3
XP_005251033.1
4
813
XM_005250978.3
XP_005251035.1
4
338
XM_005250979.4
XP_005251036.1
4
297
XM_005250980.4
XP_005251037.1
4
382
XM_005250981.3
XP_005251038.1
4
219
XM_005250982.3
XP_005251039.1
4
212
XM_005250983.2
XP_005251040.1
3
136
XM_005250984.4
XP_005251041.1
4
201
XM_006716588.3
XP_006716651.1
5
390
XM_006716589.2
XP_006716652.1
4
151
XM_006716590.3
XP_006716653.1
4
261
XM_011517130.2
XP_011515432.1
4
338
XM_011517131.2
XP_011515433.1
3
245
XM_011517132.2
XP_011515434.1
4
334
GenBank mRNA
AY480044.1
-
3
277
AY480045.1
-
3
148
AY480046.1
-
3
273
AY480047.1
-
3
771
AY480048.1
-
3
168
AY480049.1
-
3
197
AY480050.1
-
3
211
AY480051.1
-
3
319
BP231761.1
-
3
258
BP312608.1
-
4
320
BP315878.1
-
3
206
BP321327.1
-
3
205
U53204.1
-
4
322
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_201388, NM_201389, NM_201393, NM_011117, NM_001163549, and others.
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
hemidesmosome assembly
cell-cell adhesion
actin binding
protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0