Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029572.2 | NP_083848.1 | 10, 11 | 1165 | ||
GenBank mRNA | AK003217.1 | - | 10, 11 | 1134 | ||
AK141377.1 | - | 10, 11 | 1123 | |||
AK151497.1 | - | 10, 11 | 1166 | |||
AK160113.1 | - | 10, 11 | 1148 | |||
AK172973.1 | - | 1107 | ||||
AK206483.1 | - | 1, 2 | 143 | |||
AK214712.1 | - | 1, 2 | 129 | |||
BC019558.1 | - | 10, 11 | 1072 | 4192664 |