Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000329.2 | NP_000320.1 | 3 | 285 | ||
GenBank mRNA | AK289925.1 | - | 3 | 276 | ||
BC075035.2 | - | 3 | 277 | 30915627 | ||
BC075036.2 | - | 3 | 280 | 30915421 | ||
U18991.1 | - | 3 | 285 |